Evolution® Medium-Height Workstations, Kewaunee®
Supplier: Kewaunee
V407314NTK V407315NTK V407316NTK V407318NTK
V407314NTKEA 3430.13 CAD
V407314NTK V407315NTK V407316NTK V407318NTK
Evolution® Medium-Height Workstations, Kewaunee®
Furniture Tables and Workbenches
With a maximum height of 91.4 cm (36"), the starter workbenches are perfect for technical work.
  • Features a front radius edge worksurface
Starter workbenches come with two support columns, a front apron, a modesty panel, and all hardware. Add-on workstation includes all items noted above except one supporting column and base; this workstation shares the column and base from an existing starter workbench. Worksurfaces are available in black epoxy resin, ESD almond static-dissipative laminate with grounding kit, and wallaby plastic laminate.