DuPont™ Tyvek® 400 Coveralls with Respirator Hood and Attached Skid Resistant Boots, Comfort Fit Design
Supplier: DuPont
TY122SWHLG002500 TY122SWH2X002500 TY122SWH3X002500 TY122SWH4X002500 TY122SWHMD002500 TY122SWH7X002500
CA12000-570CS 1547.52 CAD
CA12000-570 CA12000-572 CA12000-574 CA12000-576 CA12000-578 CA12000-568 10016-698
DuPont™ Tyvek® 400 Coveralls with Respirator Hood and Attached Skid Resistant Boots, Comfort Fit Design
Disposable Garments
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Comfort fit design
  • Respirator fit hood
  • Zipper front
  • Elastic wrists
  • Attached skid-resistant boots
  • Elastic waist
  • Serged seams
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates toMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Gender
Garment application Laboratory