DuPont™ Tychem® 2000 Coveralls with Standard Hood and Elastic Wrists and Ankles, Bound Seams
Supplier: DuPont
Tychem®
QC127BYL2X00 QC127BYLXL00 QC127BYLMD00 QC127BYL5X001200 QC127BYLMD00 QC127BYLLG00 QC127BYLXL00 QC127BYL4X00 QC127BYL4X00 QC127BYL6X001200 QC127BYLLG00 QC127BYL3X00 QC127BYL2X00 QC127BYL3X00
CA10010-252CS 1318.82 CAD
CA10010-252 10010-252 10010-262 CA10010-254 10010-256 CA10010-260 10010-254 CA10010-262 10016-404 10016-406 10010-258 CA10010-256 CA10010-258 10010-260
DuPont™ Tychem® 2000 Coveralls with Standard Hood and Elastic Wrists and Ankles, Bound Seams
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Standard fit hood
  • Elastic wrists and ankles
  • Storm flap with adhesive closure
  • Bound seams
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration. Attached hood with elastic around face opening is standard fit. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper. Elastic openings at wrists and ankles allow for a tighter fit.
Tychem® QC coveralls provide a minimum of 30 minutes of protection against 42 chemical challenges. Coveralls are available in three seam styles; serged, bound, or taped. Garments with serged seams feature multiple interlocking threads sewn around the raw edges of the garment material to create a strong, stress-resistant seam. Garments with bound seams feature a tightly sewn seam covered with garment fabric to reinforce the seam and reduce the potential for particle penetration. Garments with bound seams also feature a storm flap covering the front zipper that can be sealed with an adhesive strip to further prevent intrusion. Garments with taped seams feature a sewn seam covered with a strip of compatible chemical-resistant material through heatsealing. Taped seam coveralls also feature a storm flap covering the front zipper that can be sealed with an adhesive strip to further prevent intrusion.
All coveralls are equipped with a standard fit hood with elastic around the face opening, and elastic wrists and ankles for a tighter fit.
The result is a rugged and tough barrier fabric that resists punctures and tears. Tychem® QC coveralls are suitable for a wide variety of applications, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. In addition, the material also provides excellent resistance against biohazards such as blood, body fluid, and viral contaminants. The highly visible yellow finish aids in worker safety by ensuring contrast across a wide range of natural backgrounds, and within both dim and brightly lit environments.
Certifications: Thickness meets ASTM D1777.