KIMBLE® KONTES® Friedrich Condensers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
437000-2440 456250-0022 437000-2942
KT437000-2440EA 807.24 CAD
KT437000-2440 KT456250-0022 KT456250-0021 KT456250-0023 10000-504
KIMBLE® KONTES® Friedrich Condensers, DWK Life Sciences
Condensers
These Friedrich condenser specially designed to provide a long vapor path, good heat transfer and anti-flooding
characteristics, used primarily in reflux mode and Soxhlet extractions.
  • Water inlet and outlet located above ring-sealed inner condensing tube
  • Molded inner spiral provides surface area for the condensation of the product
Condenser KT437000-2440, KT437000-2942 inclined standard taper outer joint on the side and standard taper inner drip joint at the bottom.
Condenser KT456250-0022, KT456250-0021, KT456250-0023 features standard taper inner drip joint and side outlet for drying tube and more.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.