Reflux Condensers, Chemglass
Supplier: Chemglass
CG-1213-05 CG-1213-06 CG-1213-01 CG-1213-03 CG-1213-07 CG-1213-09 CG-1213-14 CG-1213-19
80067-282EA 844.18 CAD
80067-282 80067-284 80067-274 80067-278 80067-286 80067-290 80067-292 80067-294
Reflux Condensers, Chemglass
Condensers
Coil style reflux condenser with tightly-wrapped coil providing maximum cooling
Excellent for use with high vapor pressure solvents. Condenser has a top outer [ST] joint and lower inner drip tip joint. Hose connections have an O.D. of 10 mm at the middle serration.