Alliance Sterile Coveralls, HPK Industries
Supplier: HPK Industries
151-S 151-XXL 151-M 151-L 151-XL
10832-160CS 2524.84 CAD
10832-160 10832-162 10832-176 10832-178 10832-180
Alliance Sterile Coveralls, HPK Industries
Disposable Garments
An extremely smooth outer fabric surface lowers particle cling and build-up in cleanroom applications
Fabric is wrinkle-free and highly durable. Built-in “cooling system” dissipates heat and moisture for high worker comfort. Elastic wrists and ankles. Bound seam construction. Sterile. Hood and boot covers (shown) not included.
Individually packaged in a sealed bag.
Coverall material acts as a liquidproof barrier. Unique antistatic system assures static dissipation in sensitive cleanroom work areas.