Snap-On Stoppers, WHEATON®
Supplier: DWK Life Sciences (KIMBLE)
224100-180 224100-080 W224100-081 W224100-181
16171-660CS 835.07 CAD
16171-660 16171-672 16171-674 89107-782
Snap-On Stoppers, WHEATON®
Closures Chromatography Vial Closures
These snap-on stoppers provide maximum sealing for vials with a blow-back glass finish, which is a small ridge on the interior of the vials neck.
  • Not recommended for vials with a straight-walled finish
The blow-back feature is typically found in vials from North American manufacturers.
The snap-on stopper locks into the blow-back ridge, providing a tight seal and keeping the stopper from backing out.