5000 Series Disposable Respirators, 3M™
Supplier: 3M
70070314409 52P71 53P71 70070314524 70070314425 70070797199 51P71 5203 7000126025 5303 5201 5101 70070797181 7000126027 70070314482 70070314466 70070314540 70070797173
CA56222-408CS 420.48 CAD
CA56222-408 CA56222-409 56222-408 56222-409 CA56222-422 CA56222-420 CA56222-464 56222-422 CA56222-410 CA56222-421 56222-460 56222-464 56222-420 CA56222-462 56222-421 56222-410 56222-462 CA56222-460
5000 Series Disposable Respirators, 3M™
Respirators Disposable Respirators Particulate Respirators
5000 series dual cartridge respirators are designed for maintenance-free, economical worker protection. Preassembled respirators are lightweight and well-balanced for high comfort. Unique cradle suspension holds respirator in place with minimal pressure to the back of the neck or bridge of the nose. Low-profile facepiece/cartridge provides a wider field of vision and greater comfort while wearing glasses or goggles. Color-coded by size. Resealable storage bag keeps respirator clean between uses.
  • Low-profile facepiece/cartridge for wider field of vision, greater comfort while wearing glasses or goggles
  • Color coded by size
  • Respirators suitable for gas and vapor protection come with replaceable prefilters
  • NIOSH approved
Respirators suitable for gas and vapor protection come with replaceable prefilters
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Respirator with P95 assembly comes with one pair of P95 prefilters and one pair of filter retainers.
Certifications: NIOSH approved.