VWR® C-Shaped Lead Ring Flask Weights
Supplier: VWR
VWR®
18308-6400 18308-6500 18308-6100 18308-6200
47735-598EA 108.69 CAD
47735-598 47735-600 47735-594 47735-596
VWR® C-Shaped Lead Ring Flask Weights
Weights Flask Weights
C-shaped lead ring flask weights are coated with green Vikem® vinyl to protect glassware.
  • Stabilize vessels in water baths or on the benchtop
  • Open ring; flexible enough to be pulled apart to accommodate a wider vessel
  • Available in four diameter/weight combinations
Flask weights fit neatly around the neck of filtering flasks with side-arm tubing connections without disconnecting the tubing.