KONTES® U-Shaped Spargers, Purge and Trap, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
591100-0005 591100-0025 591100-2605 591100-2625
KT591100-0005EA 345.15 CAD
KT591100-0005 KT591100-0025 KT591100-2605 KT591100-2625
KONTES® U-Shaped Spargers, Purge and Trap, Kimble Chase
Stirrers Stirring Tools
Designed for use with Tekmar® 2000 and 3000 purge and trap concentrators and with the 2016 and 2032 autosamplers
Fritted spargers are recommended for potable water and wastewater samples; fritless spargers are recommended for water, soils, polymers, pharmaceuticals, and foods. Mount: 1.3 cm (1/2").