ProFlex® 500 Elbow Supports, Ergodyne®
Supplier: Ergodyne
16002 16003 16004
89135-620CS 1558.4 CAD
89135-620 56618-264 56618-266
ProFlex® 500 Elbow Supports, Ergodyne®
Ergonomic Supports Elbow Supports
Supports help reduce the risk of injuries associated with repetitive upper arm movements and strong, forceful grips
They provide a wide base of support, directing pressure away from tendons of the forearm muscles. Lightweight, nylon-laminated neoprene sleeve conforms to the forearm for a comfortable fit. May be loosened when not in use. Fully-adjustable cinch strap allows exact fit and desired level of tension. Machine washable.
Ordering information: Small supports are designed to fit elbows 23–25cm (9–10"). Medium supports are designed to fit elbows 25–28cm (10–11"). Large supportsMore Product Information