Synthware Concentrator Tubes, Graduated, Kemtech America
Supplier: Kemtech America
C291910 C291925
60002-262EA 95.04 CAD
60002-262 60002-264
Synthware Concentrator Tubes, Graduated, Kemtech America
Concentrators
Graduated concentrator tubes for use with Kuderna-Danish evaporative set-ups.
  • Heavy wall Borosilicate construction
  • Graduated
  • Ground glass joint
Tube can be stoppered for storage. [ST] joint size: 19/22.
Order Now
SPECIFICATIONS
Environmentally Preferable