Positive pressure hoods provide a high level of respiratory protection while allowing easy breathing. Lightweight, loose-fitting Tychem® QC hoods feature a unique airflow design to prevent fogging. Hoods feature a solvent-resistant polyester face shield that is U-shaped to provide excellent peripheral and downward vision. Hoods are available with hard hat and collar.
- High level of respiratory protection while allowing easy breathing
- Lightweight, loose-fitting Tychem® QC feature unique airflow design to prevent fogging
- Solvent-resistant polyester face shield that is U-shaped to provide excellent peripheral and downward vision
- Available with hard hat and collar
Breathing tubes are supplied with connector and clamps
PAPR systems come with either a web belt, a vinyl belt, or a comfort belt.
Supplied air is provided through either a high- or low-pressure assembly. High-pressure assembly has three control devices to choose from. Vortex cooling assemblies let the worker regulate airflow and cool air by as much as 28°C (50°F). Vortemp™ heating assembly lets the worker regulate airflow and warm air by as much as 28°C (50°F). Air regulating valve assembly allows the worker to regulate airflow to the headgear. High-pressure air supply hose comes in 7.6m, 15.2m, or 30.5m lengths (25', 50', or 100'). The low-pressure assembly also regulates the airflow to the headgear. Low-pressure air supply hose comes in 7.6m, 15.2m, or 30.5m (25', 50', or 100') lengths.
Reusable breathing tube 56221-388 is 91cm (36") long.
Certifications: All systems are NIOSH approved when properly assembled.
Ordering information: Each PAPR system includes motor blower, battery pack, battery charger, flow meter, power cord, blower plugs, and belt. Hood assemblies, breathing tubes, and cartridges sold separately.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.