KONTES® SLOW-DRY® Concentrators, Graduated, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
570071-1025 570071-1525
KT570071-1025CS 206.03 CAD
KT570071-1025 KT570071-1525
KONTES® SLOW-DRY® Concentrators, Graduated, Kimble Chase
Concentrators
Concentrator tubes reduce the risk of concentrating a sample to dryness
Insulated air space around the constricted end of the tube shields the sample and provides a protective temperature buffer. Concentrators also feature a [ST] 19/22 top joint. All models are graduated from 0–1mL in 0.1mL divisions and in 1mL divisions from 2mL and above.
Order Now
SPECIFICATIONS
Environmentally Preferable