Alloy 8 Individual Precision Analytical Weights, Ultra Class and ANSI/ASTM Classes 1 and 4, Troemner
Supplier: Troemner
7014-1 7022-0T 7029-1T 7010-0 7035-4W 7025-1W 7012-4 7019-4T 7010-0W 7017-1W 7033-1 30391417 7027-4 7021-4 7021-1 7017-1T 7031-0 7012-4W 7013-4W 80780071 7030-1 7037-0T 7027-1W 7030-1W 7023-0W 7016-0W 7030-4W 7019-1T 7034-1W 7030-1T 7020-1 7024-4T 7036-0T 7032-4W 7010-1T 7037-4T 7034-0T 7035-1T 7033-0W 7024-1W 80780065 7023-4W 7020-4T 7017-4W 7026-1T 7013-1W 7028-1 7036-1 30391395 7025-1T 7027-0 7020-4 7033-0T 7012-0T 7031-4W 7016-0T 7037-0 7014-1T 7010-4T 7034-0W 7036-1T 7034-1T 7026-4W 7034-1 7027-0W 7010-1 80780059 7029-0W 7028-4T 7014-0W 7023-1 7026-4T 7026-4 7016-0 7036-0 7024-4 7025-4 7017-0 7036-4T 7032-0 7035-4 7032-4T 7029-0T 7031-1T 7024-4W 7035-1 7028-1W 80780073 7022-1T 7012-1 7029-0 7028-0W 7019-4 7021-4T 7012-1T 7022-0W 7022-4T 7037-1 7025-0 7010-0T 7031-4 7027-1T 7018-0 7021-0 7016-1 7021-1T 7028-1T 7032-4 7024-0W 7035-4T 7016-4 7018-1T 7016-4W 7024-0 7018-0W 7033-1W 7037-0W 7025-0T 80780060 7012-0 7034-4T 7020-4W 7019-1 7020-0 7034-0 7014-4T 7020-1T 7013-0W 7034-4W 7020-0W 7021-4W 7031-1W 7035-1W 7030-0 7029-4 7028-0T 7025-4T 7022-4 80780058 7023-1W 7026-0W 80780064 7023-0T 7014-4W 7014-1W 7032-0W 7033-4T 7017-4T 7028-4 7023-4 7036-4 7018-4 7020-1W 7035-0T 7024-1T 7019-4W 7029-1 7018-1 7031-0T 7032-0T 7024-1 7023-1T 7013-0 7017-0T 7013-1 7036-1W 7036-4W 7037-1W 7025-1 7031-0W 7029-1W 7030-4T 7016-1T 7024-0T 7018-4T 7020-0T 7031-1 7027-4W 7022-4W 7032-1T 7027-0T 7026-0 7017-1 7023-4T 7036-0W 7022-1 7012-0W 7014-4 7028-0 7037-4W 7016-1W 7012-1W 7014-0 80780066 7021-0W 7035-0 7028-4W 7022-1W 7026-0T 7027-1 7033-1T 7037-1T 7019-1W 7022-0 7033-0 7025-0W 7016-4T 7023-0 7010-4W 7030-4 80780063 7032-1 7018-0T 7029-4T 7019-0T 7013-4T 7018-4W 7033-4W 7013-4 7012-4T 7030-0T 7010-1W 7026-1 7014-0T 7037-4 7013-1T 7013-0T 7035-0W 7010-4 7021-1W 7017-0W 80780072 7031-4T 7030-0W 7027-4T 7017-4 7026-1W 7034-4 7032-1W 7018-1W 7033-4 7021-0T 7029-4W 7019-0 7025-4W 7019-0W
12764-988EA 383.58 CAD
AP400006-910 12764-988 12764-989 12765-670 12765-550 12765-551 12765-430 12765-672 12765-552 12765-553 12765-432 12765-674 12765-554 12765-676 12765-434 12765-555 12765-678 12765-436 12765-668 12765-426 12765-547 12765-548 12765-428 12765-549 12764-990 12764-993 12764-994 12764-991 12764-992 12764-995 12764-979 12766-588 12764-977 12766-586 12764-978 12766-584 12766-582 12765-660 12766-580 12765-540 12765-420 12765-662 12765-541 12765-542 12765-422 12765-664 12765-543 12765-544 12765-424 12765-545 12765-546 12765-536 12765-537 12765-416 12765-658 12765-538 12765-539 12765-418 12764-982 12764-983 12764-980 12764-981 12764-986 12764-987 12764-984 12764-985 12765-690 12765-692 12765-694 12765-696 12765-698 12765-680 12765-682 12765-440 12765-684 12765-686 12765-688 12765-438 12766-554 12766-552 12766-550 12765-630 12765-510 12765-511 12765-632 12765-512 12765-634 12765-513 12765-624 12765-626 12765-628 12764-950 12766-548 12764-953 12764-954 12764-951 12766-546 12764-952 12766-544 12764-936 12766-542 12764-934 12766-540 12764-938 73690-038 73690-036 12765-620 12765-622 12765-614 12765-616 12765-618 12766-538 12764-940 12766-536 12764-968 12766-578 12764-969 12764-966 12764-967 12766-576 12766-574 73690-048 12766-572 73690-046 12765-650 12766-570 73690-044 12765-530 12765-652 12765-410 73690-042 12765-654 12765-412 73690-040 12765-535 12765-656 12765-414 12765-404 12765-525 12765-646 12765-526 12765-527 12765-648 12765-406 12765-528 12765-529 12765-408 12764-970 12764-975 12764-976 12766-568 12766-566 12764-957 12764-958 12766-564 12764-955 12764-956 12755-556 12766-562 12764-959 12766-560 12765-640 12765-520 12765-521 12765-400 12765-642 73690-054 12765-522 12765-523 12765-644 12765-402 12765-524 73690-052 12765-514 12765-515 73690-050 12765-636 12765-516 12765-517 12765-638 12765-518 AP400006-884 12765-519 12764-960 12764-961 12764-964 12764-965 12764-962 12764-963 12766-558 12764-902 12766-512 12764-900 12766-510 12764-906 12764-904 12764-908 12765-710 12765-702 12765-704 12765-706 12765-708 12766-508 12766-506 12766-504 12764-910 12766-502 12766-500 12765-700 12764-924 12766-534 12764-922 12766-532 12764-928 12766-530 12764-926 12765-610 12765-612 12765-602 12765-724 12765-604 12765-726 12765-606 12765-608 12766-528 12766-526 12764-932 12766-524 12764-930 12766-522 12764-914 12766-520 12764-912 12764-918 12764-916 12765-720 12765-600 12765-722 12765-712 12765-714 12765-716 12765-718 12766-518 12764-920 12766-516 12766-514
Alloy 8 Individual Precision Analytical Weights, Ultra Class and ANSI/ASTM Classes 1 and 4, Troemner
Weights Calibration Weights
Individual weights available in Troemner UltraClass (equals or exceeds OIML Class E2 tolerances), and ANSI/ASTM Classes 1 and 4.
Fractional weights 500mg to 5mg are made of stainless steel (30mg weights are aluminum); fractional weights 3mg and below are made of aluminum.
UltraClass weights 1g to 20kg are made from Troemner Alloy 8, which is a specially austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability and resistance to corrosion and handling damage. This exclusive material has a density of 8.0 at 25ºC (controlled to ±0.1%), is essentially non-magnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to –195.5ºC (–320ºF). Class 1 and 4 weights 1g to 1kg are made from Troemner Blended Stainless Steel. Weights 2kg to 50kg are made from Type 316 stainless steel.
Certifications: Where applicable, weights are supplied with a NIST/NVLAP Weight Calibration Certificate or a Traceable Certificate.
Ordering information: For ANSI/ASTM Class 2 and Class 3 analytical weights, contact your Avantor sales representative.
Delivery information: Weights 12765-632 (30390176) and 12765-640 (30390172) are supplied without certification.