Nalgene® Laboratory Notebooks, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
6301-4000 6301-1000 6301-3000 6301-2000
51280-110CS 647.14 CAD
51280-110 51280-112 28196-344 51280-108
Nalgene® Laboratory Notebooks, Thermo Scientific
Notebooks
Notebooks feature sewn-in paper pages with either horizontal ruled lines or grid
Pages have spaces for dates, signatures of workers and witnesses, and other necessary information for securing patent protection. Polyethylene cover is waterproof. Notebooks have 100 numbered pages.