KLEENGUARD® A40 Liquid and Particle Protection Shoe Covers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Microflex
44490 44492 44494
CA82007-666CS 401.17 CAD
CA82007-666 CA82007-668 CA82007-670
KLEENGUARD® A40 Liquid and Particle Protection Shoe Covers, KIMBERLY-CLARK PROFESSIONAL®
Disposable Garments
Shoe covers provide an excellent liquid and particle barrier. They are designed to be used in applications where Tyvek® material is typically worn. KLEENGUARD® A40 is a lamination of microporous film and a spunbond nonwoven material. KLEENGUARD® A40 laminate has a dry particle holdout of 100% between 0.3–0.5µm.
  • Used in applications where Tyvek® material is typically worn
  • Lamination of microporous film and spunbond nonwoven material
  • Dry particle holdout of 100% between 0.3–0.5µm
Shoe covers have elastic at the tops
Meet ANSI/ISEA 101-1996.
Universal shoe cover 82007-666 has an elastic seam across the bottom and a length of 41.9cm (161/2") when stretched. Large and X-large shoe covers (82007-668 and -670) feature a sewn-in vinyl sole for traction and durability. The length from tip of toe to heel is 33.7cm (131/4") for 82007-668 and 38.1cm (15") for 82007-670.