VWR® Aluminum Micro Weighing Dishes
Supplier: VWR International
MAWD-0350 MAWD-3500 MAWD-0400 MAWD-1100 MAWD-1120 MAWD-1200 MAWD-1201
12577-060PK 236.78 CAD
12577-060 12577-062 12577-064 12577-066 12577-068 12577-070 12577-072
VWR® Aluminum Micro Weighing Dishes
Boats Weighing Boats
Ideal for microbalances, organic micro elementary and thermal analysis, and pharmaceutical use.
  • Easy to use
  • Compatible with most commercially available analyzers and microbalances
  • For use in a variety of applications
Our micro weighing boats are compatible with most commercially available analyzers and microbalances. Packaged in easy-to-use plastic containers for storage and dispensing.