Dissecting Board and Base, Simport Scientific
Supplier: Simport Scientific
M625 M620
CA89041-172EA 290.58 CAD
CA89041-172 CA89041-170
Dissecting Board and Base, Simport Scientific
Dissecting Boards
Stain-resistant, two-sided dissecting board is constructed of heavy-duty polyethylene
The lightweight, non-skid dissecting board base is used to elevate the dissecting board to the appropriate height. The base can contain excess fluids and is stackable for various height requirements.
One side features squares, circles, and dimensional scales in inches and centimeters. The other side features two cutting board sections with half-size dimensional scales. Dimensions: 61Lx43.2W cm (24x17"). Weight: 3.1 kg (6.8 lbs.).
A smaller DissecTable™ is also available with the same features and benefits as the 89041-170. Perfect for smaller counter areas. Dimensions: 33.0×27.9×1.25 cm (13×11×0.5").