Stainless Steel Metric Weight Sets, Class F, Troemner
Supplier: Troemner
30390418 30390400 30390411 30390406
12765-480EA 851.28 CAD
12765-480 12765-485 12765-490 12765-495
Stainless Steel Metric Weight Sets, Class F, Troemner
Weights Calibration Weights
Used primarily to test commercial weighing devices
Sets are supplied in a durable polypropylene case with padded handle and fitted foam interior.
Precision-crafted stainless steel weights are calibrated according to NIST specifications for Class F. NIST Class F is defined by NIST Handbook 105-1, which specifies weights used by government officials to verify commercial weighing devices. Class F is also acceptable for use in verifying scales that have a 0.1% accuracy or lower.