DuPont™ Tyvek® 400 Frocks with Laydown Collar
Supplier: DuPont
Tyvek®
TY210SWHXL00 TY210SWH3X00 TY210SWH3X00 TY210SWH2X00 TY210SWH5X003000 TY210SWHMD00 TY210SWHSM00 TY210SWHSM00 TY210SWH7X003000 TY210SWH2X00 TY210SWHMD0030VP TY210SWHLG00 TY210SWHXL00 TY210SWH2X0030VP TY210SWHXL0030VP TY210SWHLG0030VP TY210SWH4X00 TY210SWHLG00 TY210SWH3X0030VP TY210SWH4X00 TY210SWH6X003000 TY210SWH4X0030VP TY210SWHMD00
CA10010-076CS 1425.2 CAD
CA10010-076 10010-076 10016-918 CA10010-078 CA10010-068 10010-074 CA10010-072 76188-104 CA10010-074 10010-068 76188-102 10010-078 76188-312 76188-100 10016-920 76188-310 10016-922 76188-098 10010-080 CA10010-080 CA10010-070 10010-072 10010-070
DuPont™ Tyvek® 400 Frocks with Laydown Collar
Disposable Garments
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Laydown collar
  • Open wrists
  • Extends below hips
  • Front snap closure
  • Serged seams
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Frocks feature body-length jacket design with front snap closures for easy donning and doffing. Ideal for applications that do not require the full body coverage of a coverall. Frocks also feature a laydown collar.
Certifications: Thickness meets ASTM D1777.