KLEENGUARD® A40 Liquid and Particle Protection Lab Coats, Kimberly-Clark Professional®
Supplier: Microflex
44442 44443 44445 44446 44452 44453 44454 44455 44456 44444
CA82007-616CS 299.4 CAD
CA82007-616 CA82007-618 CA82007-622 CA82007-624 CA82007-626 CA82007-628 CA82007-630 CA82007-632 CA82007-634 CA82007-620
KLEENGUARD® A40 Liquid and Particle Protection Lab Coats, Kimberly-Clark Professional®
Disposable Garments
Manufactured from a breathable microporous film laminate, lab coats provide an excellent particle barrier and resist nonhazardous liquids
Meets ANSI/ISEA 101-1996. Passes ASTM F1670 testing for penetration of blood and body fluids. NFPA compliant antistatic material.
These low-linting garments are ideally suited for use in manufacturing, cleaning, utility, liquid handling, pressure washing, and maintenance applications. The tough exterior film protects while its soft nonwoven fabric interior provides comfort against the skin. KLEENGUARD® A40 laminate has a dry particle holdout of 100% between 0.3–0.5µm.