Synthware Vacuum Take-Off Adapters, Short Stem, Kemtech America
Supplier: Kemtech America
A621420 A621922 A622440 A622942
60000-634EA 101.92 CAD
60000-634 60000-636 60000-638 60000-640
Synthware Vacuum Take-Off Adapters, Short Stem, Kemtech America
Adapters Adapters, Joints with Tube Connectors
These vacuum take-off adapters have a short stem.
  • Top outer [ST] joint
  • Bottom inner [ST] joint
  • With hose connection and drip tip
Featuring a hose connection and drip tip. Hose connection has an 8 mm ext. Ø. The top joint is outer [ST] and the bottom joint is inner [ST].