VWR® Gooch Crucibles, Porcelain
Supplier: VWR International
89038-038 89038-040 89038-042 89038-044 89038-046
89038-038CS 472.39 CAD
89038-038 89038-040 89038-042 89038-044 89038-046
VWR® Gooch Crucibles, Porcelain
Crucibles
Chemical-porcelain gooch crucibles are used in total suspended solids determinations.
  • Glazed interior and exterior
  • Chemical-resistant
Glazed inside and out except outside bottom surface and rim. Perforated bottom. The Gooch Crucible with Wall Holes has two holes for suspension in an extraction apparatus. For use in temperatures up to 1150°C (2102°F).
Ordering information: Covers are sold separately.