Chemware® Teflon® Resin Evaporating Dishes, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
D1069044 D1069045
25337-059EA 138.47 CAD
25337-059 25337-070
Chemware® Teflon® Resin Evaporating Dishes, Saint-Gobain Performance Plastics
Trays/Dishes/Pots Evaporating Basins/Dishes
With broad pouring spouts and flat bottoms
These resin evaporating dishes, saint-gobain performance plastics are made of PTFE.
Chemically inert to virtually all materials. Heat resistant to 260°C (550°F).
Order Now
SPECIFICATIONS
Environmentally Preferable