Adapters with PTFE Plug, Ace Glass Incorporated
Supplier: Ace Glass
5202-92 5202-12 9080-12 9080-18 9080-112 9080-118 5202-110 5202-112 5202-114
71001-508EA 282.03 CAD
71001-508 89051-398 89064-854 89064-856 89082-708 89082-710 89082-736 89082-764 89082-770
Adapters with PTFE Plug, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Adapters feature either a 90° angled or straight hose connection, with ground [ST] inner joint at bottom and solid PTFE stopcock plug.
  • 90° angled or straight hose connection
  • Bottom ground [ST] inner joint
  • Solid PTFE stopcock plug
  • Manufactured from borosilicate glass
Featuring a 1:5 solid PTFE 2mm bore stopcock plug.
Ordering information: For replacement stopcock plug, see 89062-770 series.
Order Now
SPECIFICATIONS
Environmentally Preferable