KIMBLE® KONTES® Inlet Adapters, with Hose Connection, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
276760-0000 205000-2440
KT276760-0000EA 232.65 CAD
KT276760-0000 KT205000-2440 KT205000-2942 KT276750-0000
KIMBLE® KONTES® Inlet Adapters, with Hose Connection, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Vertical adapters with vacuum take-off and drip tip.
  • Top outer [ST] joint
  • Bottom inner [ST] joint
  • Both joints are same size
  • With hose connection and drip tip
The outer Standard Taper joint at the top and inner Standard Taper joint at the bottom are the same size.