Claisen Adapters, Ace Glass Incorporated
Supplier: Ace Glass
5135-10 5135-06 5135-08 5135-12 5135-14
89051-254EA 364.56 CAD
89051-254 89084-940 89084-944 89084-948 89084-952
Claisen Adapters, Ace Glass Incorporated
Distillation Components Distillation Adapters
Adapters feature a top outer [ST] joint, bottom inner [ST] joint, and side inner [ST] joint
  • Top outer [ST] joint
  • Bottom inner [ST] joint
  • Side inner [ST] joint
  • Side tube accommodates thermometer
  • Manufactured from borosilicate glass
The top of the side tube has a [ST] 10/30 joint for a 76mm (3") immersion thermometer.
The top of the side tube accommodates a thermometer.