SP Bel-Art Antibody Saver Tray, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F45100-0000 F45100-0001 F45100-0002
89042-158CS 486.12 CAD
89042-158 89047-216 97000-900
SP Bel-Art Antibody Saver Tray, Bel-Art Products, a part of SP
Trays/Dishes/Pots Spillage Trays/Pans
Minimize the use of expensive antibodies during incubations by using an optimally sized tray.
  • Stackable
  • Polypropylene construction
  • Attached lid protects against spillage
  • Autoclavable
These polypropylene trays are available for popular midi and mini gels and include an attached lid to protect against spillage. Single lane trays are supplied as a group of five joined lanes that can be easily separated to single lane prior to use. Trays are stackable, reclosable, and reusable.