Synthware Anti-Splash Adapters
Supplier: Kemtech America
A282414 A282419 A282424 A282914 A282924 A281250 A289250 A284250 A282250 A288250
60000-408EA 228.49 CAD
60000-408 60000-410 60000-412 60000-414 60000-418 60000-422 60000-424 60000-426 60000-428 60000-430
Synthware Anti-Splash Adapters
Adapters Adapters, Conical Ground Joints
Anti-splash adapters are designed to prevent possible contamination from a rotary evaporator.
  • Top outer [ST] joint
  • Bottom inner [ST] joint
The upper joint is an outer [ST] joint, while the lower joint is an inner [ST] joint.