Synthware Enlarging Adapters, Kemtech America
Supplier: Kemtech America
A251914 A252414 A252419 A252914 A252924 A253424 A254024 A254529
60000-052EA 60.98 CAD
60000-052 60000-056 60000-058 60000-060 60000-062 60000-064 60000-066 60000-070
Synthware Enlarging Adapters, Kemtech America
Adapters Adapters, Conical Ground Joints
Enlarging adapters are useful for attaching apparatus with one joint size to equipment having a larger joint size.
  • Top outer [ST] joint
  • Bottom inner [ST] joint
Features an outer [ST] joint at the top and inner [ST] joint at the bottom.