Bushing Adapters, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
13430-05 13430-07 13430-11 13430-13 13430-16 13430-18 13430-21 13430-25 13430-28 13430-32 13430-40 13430-42 13430-44
89082-672EA 72.46 CAD
89082-672 89082-674 89082-676 89082-678 89082-680 89082-682 89082-684 89082-686 89082-688 89082-690 89085-126 89085-128 89085-130
Bushing Adapters, PTFE, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
These bushing-type adapters are manufactured from polytetrafluoroethylene.
  • Top inner [ST] joint
  • Bottom outer [ST] joint
Adapters feature an inner [ST] joint at the top, and an outer [ST] joint at the bottom.