Adapters, Enlarging, Ace Glass Incorporated
Supplier: Ace Glass
5005-06 5005-10 5005-12 5005-14 5005-24 5005-28 5005-36 5005-38 5005-42 9092-10 9092-20 9092-21 9092-22 9092-26 9092-27 9092-28 5005-23 5005-30 5005-08
89051-014EA 111.85 CAD
89051-014 89051-018 89051-020 89051-022 89051-024 89051-026 89051-028 89051-030 89051-032 89064-874 89064-882 89064-884 89064-886 89064-892 89064-894 89064-896 89082-560 89082-568 89051-016
Adapters, Enlarging, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
These enlarging adapters are manufactured from borosilicate glass.
  • Larger top [ST] joint
  • Smaller bottom [ST] joint
Adapters feature standard taper joints at both top and bottom. The top joint is larger than the bottom joint. All full-length outer standard taper joints are reinforced.
Order Now
SPECIFICATIONS
Environmentally Preferable