SP Bel-Art Large Capacity Multipurpose Trays, Polypropylene, Bel-Art, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F16200-0010 F16200-0011
74950-086EA 96.09 CAD
74950-086 74950-088
SP Bel-Art Large Capacity Multipurpose Trays, Polypropylene, Bel-Art, Bel-Art Products, a part of SP
Trays/Dishes/Pots Laboratory Trays/Dishes
Rugged, large capacity 8.5L (9qt.) trays may be used to contain ice, a water bath, glassware, or instruments for autoclaving.
  • Rugged durability
  • Polypropylene construction
  • Available with or without handles
  • Autoclavable
Inside corners are smooth and rounded for easy cleaning. Top rim adds rigidity and provides handholds for transport. Identical trays can nest for efficient storage. Trays are available with or without handles. Steam autoclavable at 121°C (250°F).