KIMBLE® KONTES® Reducing Adapters, Bushing Style, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
150750-0242 150750-0223 150750-0265 150750-0266 150750-0268 273500-0022
KT150750-0242EA 151.57 CAD
KT150750-0242 KT150750-0223 KT150750-0265 KT150750-0266 KT150750-0268 KT273500-0022
KIMBLE® KONTES® Reducing Adapters, Bushing Style, DWK Life Sciences
Adapters Adapters, Conical Ground Joints
Reducing adapters allow for compact [ST] assembly of dissimilar joints.
  • Bushing style
  • Top and bottom [ST] joints
Adapters with a [ST] 10/30 top joint are useful as fittings for [ST] thermometers.
Order Now
SPECIFICATIONS
Environmentally Preferable