Bushing Adapters, Chemglass
Supplier: Chemglass
CG-1016-12 CG-1016-T-02 CG-1016-T-03 CG-1016-T-05 CG-1016-T-06 CG-1016-04 CG-1016-05 CG-1016-06 CG-1016-10 CG-1016-13 CG-1016-17 CG-1016-18 CG-1016-19
80065-764EA 165.2 CAD
80065-764 80060-268 80060-270 80060-274 80060-276 80065-750 80065-752 80065-754 80065-762 80065-766 80065-772 80065-774 80065-776
Bushing Adapters, Chemglass
Adapters Adapters, Conical Ground Joints
These bushing-style adapters are available in borosilicate glass or fluoropolymer resin.
  • Lower [ST] inner joint
  • Outer joint ground in bore of adapter
Bushing-style adapters have a lower [ST] inner joint and an outer joint ground in the bore of the adapter. Top outer joint [ST] 14/35 can be used with 14/20 joints.
Ordering information: For additional [ST] joint sizes, contact your Avantor sales representative.