Connecting [ST] Adapters, Chemglass
Supplier: Chemglass
CG-1000-05 CG-1000-10 CG-1000-13 CG-1000-14 CG-1000-29 CG-1000-30 CG-1000-31 CG-1000-43 CG-1000-44 CG-1002-07 CG-1002-09 CG-1002-15 CG-1002-16 CG-1002-31
80065-474EA 107.88 CAD
80065-474 80065-484 80065-488 80065-490 80065-500 80065-502 80065-504 80065-522 80065-524 80065-544 80065-546 80065-552 80065-554 80065-574
Connecting [ST] Adapters, Chemglass
Adapters Adapters, Conical Ground Joints
Connecting [ST] adapters are designed to connect dissimilar [ST] joint sizes.
  • Reducing and enlarging adapters available
  • Choose from a variety of [ST] joint sizes
For reducing adapters, lower inner joint is larger in size than the top outer joint. For enlarging adapters, lower inner joint is smaller in size than the top outer joint.
Ordering information: For additional sizes, contact your Avantor sales representative.
Order Now
SPECIFICATIONS
Environmentally Preferable