KIMBLE® KONTES® Connecting [ST] Adapters for Distilling, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
167500-2440 167000-2440 274950-0000 275060-0000
KT167500-2440EA 276.69 CAD
KT167500-2440 KT167000-2440 KT274950-0000 KT275060-0000
KIMBLE® KONTES® Connecting [ST] Adapters for Distilling, DWK Life Sciences
Distillation Components Distillation Adapters
Use connecting [ST] adapter as a component in atmospheric or vacuum distillations.
  • Inner joints are at a 75° angle
  • For use in distillation assemblies
Adapter KT167000-2440 has a [ST] top joint for a thermometer.