Cleanroom Wiper Dispensers, S-Curve Technologies
Supplier: S-Curve Technologies
WD-9.5 PETG WD-12.5 PETG
32948-865EA 218.29 CAD
32948-865 32948-867 89140-770 89140-772
Cleanroom Wiper Dispensers, S-Curve Technologies
Wipe Dispensers
Wiper dispensers are made of clean, clear, 6 mm (1/4") thick PETG with flame-treated edges for a smooth finish.
  • Protective covers help maintain the cleanliness of the wipers
  • Table top only
Keep 22.9x22.9 cm (9x9") or 30.5x30.5 cm (12x12") wipers clean and easily accessible in these convenient wiper dispensers.