Spill Containment Pallet/Platforms with Grating, Eagle Manufacturing
Supplier: Eagle Manufacturing
1686 1688 1647 1645
56610-161EA 1448.64 CAD
56610-161 56610-163 56610-202 56617-333
Spill Containment Pallet/Platforms with Grating, Eagle Manufacturing
Drum Storage
Low-profile pallet and platform units support 208.2L (55gal.) drums and provide secondary spill containment.
  • Low-profile units support 208.2L (55gal.) drums
  • Provide secondary spill containment
All-polyethylene construction with excellent chemical resistance. Translucent tub is safety yellow with black grating supports. Grating has 4.4cm (13/4") square openings and removes easily for cleaning. Pallet unit 56617-333 is forkliftable and has a 1.9cm (3/4") drainplug. Ramp 56610-212 fits platform units.
Certifications: Meet EPA requirement 40 CFR 264.175.