Synthware Drying Tube Adapters, 75°, Kemtech America
Supplier: Kemtech America
A871420 A871922 A872440
60000-582EA 72.02 CAD
60000-582 60000-584 60000-586
Synthware Drying Tube Adapters, 75°, Kemtech America
Adapters Adapters, Conical Ground Joints
Drying tube adapters are used for keeping a reaction vessel free of moisture.
  • 75° angle bend
  • With joint
Adapters are angled at 75° to avoid desiccant contact with reactions.