Nalgene® Pans, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
6902-1000 6902-2000 6902-3000 6902-5000
62662-150CS 411.04 CAD
62662-150 62662-183 62662-219 62662-241
Nalgene® Pans, Polypropylene, Thermo Scientific
Trays/Dishes/Pots Laboratory Trays/Dishes
Pans are lightweight, durable, chemically inert, and autoclavable
For repeated autoclaving of small instruments and labware. Can also be used for soaking, cleaning, storing, or carrying general lab products. Heavy-duty construction with extra strong rims. Tapered for nesting.