Synthware Anti-Splash Adapters with Fritted Disc, Modified, Kemtech America
Supplier: Kemtech America
A462414 A462424 A461250 A464250
60000-504EA 274.68 CAD
60000-504 60000-508 60000-518 60000-522
Synthware Anti-Splash Adapters with Fritted Disc, Modified, Kemtech America
Adapters Adapters, Conical Ground Joints
Anti-splash adapters prevent possible contamination from evaporator.
  • Two 4mm drain holes
  • Extra coarse fritted disc
Adapters feature two 4mm drain holes at the base of the inner tube to allow solvent to drain back into the evaporation flask and an extra coarse fritted disc sealed above lower inner joint. Frit reduces foam overflow from being drawn into the condenser.