Phloroglucinol, anhydrous ≥99.9%
Supplier: CHEM-IMPEX INTERNATIONAL, INC
Synonyms:
1,3,5-Trihydroxybenzene
24456-100G
24456-1KG
24456-25G
CH24456-100GEA
74.21
CAD
CH24456-100G
CH24456-1KG
CH24456-25G
Phloroglucinol, anhydrous ≥99.9%
Phloroglucinol
Formula:
C₆H₆O₃ MW: 126.11 g/mol Boiling Pt: 400 Storage Temperature: Refrigerator |
MDL Number:
MFCD00002286 CAS Number: 108-73-6 Merck Index: 12,07482 |
Specification Test Results
Assay by titration | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...