PinPoint™ Pipette Controller
Supplier: United Scientific Supplies
UNPINPNT01 UNPINPNT02 UNPINPNT03 UNPINPNT04 UNPINPNT05
470356-092EA 317.57 CAD
470356-092 470356-086 470356-094 470356-088 470356-090
PinPoint™ Pipette Controller
Pipettes Pipette Controllers
Digital display makes transferring liquids easier.
  • Compatible with glass or plastic serological pipettes (0.1 to 100 ml)
  • LCD indicates battery charge and 6 speed levels
  • Fills a 25 ml pipette in less than 5 seconds
  • Lithium-ion battery charges fully within 3 hours and can provide up to 8 hours of use
Ultra-light and powerful pipette controller is designed for comfort as well as accuracy. Intuitive 6 level speed setting and sensitive triggers provide a wide range of aspirating/dispensing speeds. Pipette controller is ergonomically designed for extendedMore Product Information