United Scientific™ Flat Electrode Strips
Supplier: United Scientific Supplies
UNELCAL UNELCC UNELCCU UNELCFE UNELCSET UNELCTIN UNELCZN
470356-068EA 2.92 CAD
470356-068 470356-058 470356-054 470356-014 470356-030 470356-020 470356-060
United Scientific™ Flat Electrode Strips
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
A variety of electrodes for electrochemical cells.
  • Available in six different metals
Use these flat, rigid electrodes in a variety of electrochemical demonstrations and experiments. The 127 mm long strips can fit into a variety of beakers, wet cells, and porous cups. If needed, the top can be bent over to stand upright in a container. Since the strips consist of one type of metal, they can be used in electroplating demonstrations. Also available as a set of 12 electrodes (two each of aluminum, carbon, copper, iron, tin, and zinc).