SSIbio® DuraFrame® Rigid qPCR Plates with White Wells, Scientific Specialties
Supplier: SCIENTIFIC SPECIALTIES, INC.
3735TW0S 3735-W0S 3742TW0S 3742-W0S
77506-834CS 1199.57 CAD
77506-834 77506-836 77506-854 77506-856
SSIbio® DuraFrame® Rigid qPCR Plates with White Wells, Scientific Specialties
Microplates PCR Plates
DuraFrame® plates feature a rugged polycarbonate frame, providing a solid and flat surface that guarantees seamless interaction with automated liquid handling platforms. With a firm grip assured, robots can confidently place DuraFrame® PCR plates with the utmost precision, eliminating any concerns of misalignment.
  • Polycarbonate frame is rigid and flat to facilitate a firm grip by robotic arms and ensure reliable placement
  • Polypropylene wells provide optimal thermal transfer and higher reaction efficiency
  • White wells enhance the fluorescence signal during qPCR
  • Wells are slightly raised to ensure evaporation-proof sealing with film, foil, or strip caps
  • ANSI/SBS footprint and well spacing
  • Vibrant colors offer a visual distinction between different samples or experiments
DuraFrame® plates feature polypropylene wells to facilitate optimal thermal transfer and higher reaction efficiency. This translates to more accurate and reliable results, giving researchers the confidence they need in their experiments. Furthermore, white well options have been meticulously crafted to enhance the fluorescence signal during qPCR, enabling clearer and more precise measurements.
Sealing PCR plates is a crucial aspect of laboratory workflows, and DuraFrame® excels in this domain as well. The slightly raised wells of DuraFrame® PCR plates guarantee an evaporation-proof seal with film, foil, or strip caps. No more worries about sample integrity or loss due to evaporation, DuraFrame® ensures that valuable samples are protected throughout the entire process.
With an ANSI/SBS footprint and well spacing, DuraFrame® PCR plates seamlessly integrate into existing laboratory setups, offering compatibility and ease of use. Moreover, the vibrant color options provide a visual distinction between different samples or experiments, allowing quick and efficient identification and organization.
For plates with clear polypropylene wells, see item 77506-838.
Certifications: Free of DNase, RNase, DNA, and PCR inhibitors.
Packaging: Plates are packaged 10 per pack, 10 packs per case.