AccuForge® Contour Blades
Supplier: Accutec Blades
AGBL-9000-0000 AGBL-9001-0000
77322-516CS 1747.2 CAD
77322-516 77322-518
AccuForge® Contour Blades
Razors and Blades
AccuForge® contour blades feature precision ground edges for high accuracy cutting applications.
  • Great for precision cuts
  • Degreased to remove oil residue
  • Available in standard or long point
  • High-quality stainless steel
  • Made in USA
Slot-in blade allows attachment to #3 handles.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.