AccuForge® Single Edge Blades, Unwrapped
Supplier: Accutec Blades
AGBL-7025-0000 AGBL-7047-0000 AGBL-7005-0000 AGBL-7038-0000
76780-338CS 4093.94 CAD
76780-338 77322-506 77322-466 77322-496 77322-500
AccuForge® Single Edge Blades, Unwrapped
Razors and Blades
AccuForge® Single Edge blades feature precision ground edges for sharpness and durability.
  • Bulk, unwrapped blades are ideal for high volume use
  • Choose 3-facet blades for increased sharpness, or 2-facet blades for increased durability
  • Available in 0.012" heavy-duty carbon steel
  • Available in No-Notch design
These AccuForge® blades are unwrapped for high volume use.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.