AccuForge® Single Edge Blades, 10 Blade Safety Dispenser
Supplier: Accutec Blades
AGBL-7023-0000 AGBL-7040-0000
77322-482CS 4826.99 CAD
77322-482 77322-492 77322-498
AccuForge® Single Edge Blades, 10 Blade Safety Dispenser
Razors and Blades
AccuForge® Single Edge blades feature precision ground edges for sharpness and durability.
  • Transparent safety dispenser with used blade vault
  • Choose 3-facet blades for increased sharpness, or 2-facet blades for increased durability
  • Each dispenser contains 10 blades
Each dispenser holds 10 blades and includes used blade storage.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.